missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-7-methylchromone 98.0+%, TCI America™

Catalog No. C30351G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30351G Supplier TCI America Supplier No. C30351G
Only null left

Chemical Identifiers

CAS 67029-84-9
Molecular Formula C10H7ClO2
Molecular Weight (g/mol) 194.614
MDL Number MFCD00239401
InChI Key UQXYHMICNLSDMN-UHFFFAOYSA-N
Synonym 6-chloro-7-methylchromone, 6-chloro-7-methyl-4h-chromen-4-one, 6-chloro-7-methyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one,6-chloro-7-methyl
PubChem CID 688707
IUPAC Name 6-chloro-7-methylchromen-4-one
SMILES CC1=C(C=C2C(=O)C=COC2=C1)Cl

Specifications

Melting Point 171°C
Color Yellow
Quantity 1 g
Formula Weight 194.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-7-methylchromone