missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-7-deazapurine 98.0+%, TCI America™
$214.37 - $663.60
Chemical Identifiers
| CAS | 3680-69-1 |
|---|---|
| Molecular Formula | C6H4ClN3 |
| Molecular Weight (g/mol) | 153.569 |
| MDL Number | MFCD01686865 |
| InChI Key | BPTCCCTWWAUJRK-UHFFFAOYSA-N |
| Synonym | 4-chloro-7h-pyrrolo 2,3-d pyrimidine, 4-chloropyrrolo 2,3-d pyrimidine, 6-chloro-7-deazapurine, 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro, 1h-pyrrolo 2,3-d pyrimidine, 4-chloro, 6-chloro-7-deazapurine 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-1h-pyrrol 2,3-d pyrimidine |
| PubChem CID | 5356682 |
| IUPAC Name | 4-chloro-7H-pyrrolo[2,3-d]pyrimidine |
| SMILES | C1=CNC2=C1C(=NC=N2)Cl |
Chemical Identifiers
| 3680-69-1 | |
| 153.569 | |
| BPTCCCTWWAUJRK-UHFFFAOYSA-N | |
| 5356682 | |
| C1=CNC2=C1C(=NC=N2)Cl |
| C6H4ClN3 | |
| MFCD01686865 | |
| 4-chloro-7h-pyrrolo 2,3-d pyrimidine, 4-chloropyrrolo 2,3-d pyrimidine, 6-chloro-7-deazapurine, 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro, 1h-pyrrolo 2,3-d pyrimidine, 4-chloro, 6-chloro-7-deazapurine 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-1h-pyrrol 2,3-d pyrimidine | |
| 4-chloro-7H-pyrrolo[2,3-d]pyrimidine |
Specifications
| 3680-69-1 | |
| White-Yellow | |
| MFCD01686865 | |
| 4-chloro-7h-pyrrolo 2,3-d pyrimidine, 4-chloropyrrolo 2,3-d pyrimidine, 6-chloro-7-deazapurine, 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro, 1h-pyrrolo 2,3-d pyrimidine, 4-chloro, 6-chloro-7-deazapurine 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-1h-pyrrol 2,3-d pyrimidine | |
| C1=CNC2=C1C(=NC=N2)Cl | |
| 153.569 | |
| 153.57 | |
| Crystalline Powder |
| 188°C | |
| C6H4ClN3 | |
| 1 g | |
| BPTCCCTWWAUJRK-UHFFFAOYSA-N | |
| 4-chloro-7H-pyrrolo[2,3-d]pyrimidine | |
| 5356682 | |
| ≥98.0% (GC,T) | |
| 6-Chloro-7-deazapurine |
Safety and Handling
RTECSNumber : UY9360000
TSCA : No