missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-7-deazapurine 98.0+%, TCI America™

Catalog No. C23061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23061G 1 g
C23065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23061G Supplier TCI America Supplier No. C23061G

Chemical Identifiers

CAS 3680-69-1
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.569
MDL Number MFCD01686865
InChI Key BPTCCCTWWAUJRK-UHFFFAOYSA-N
Synonym 4-chloro-7h-pyrrolo 2,3-d pyrimidine, 4-chloropyrrolo 2,3-d pyrimidine, 6-chloro-7-deazapurine, 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 7h-pyrrolo 2,3-d pyrimidine, 4-chloro, 1h-pyrrolo 2,3-d pyrimidine, 4-chloro, 6-chloro-7-deazapurine 4-chloro-1h-pyrrolo 2,3-d pyrimidine, 4-chloro-1h-pyrrol 2,3-d pyrimidine
PubChem CID 5356682
IUPAC Name 4-chloro-7H-pyrrolo[2,3-d]pyrimidine
SMILES C1=CNC2=C1C(=NC=N2)Cl

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 1 g
Formula Weight 153.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-7-deazapurine
RTECSNumber UY9360000
TSCA No