missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-5-(2-chloroethyl)oxindole 98.0+%, TCI America™
$354.23 - $354.23
Chemical Identifiers
| CAS | 118289-55-7 |
|---|---|
| Molecular Formula | C10H9Cl2NO |
| Molecular Weight (g/mol) | 230.088 |
| MDL Number | MFCD03411598 |
| InChI Key | ZTQQXEPZEYIVDK-UHFFFAOYSA-N |
| Synonym | 6-Chloro-5-(2-chloroethyl)-2-indolinone |
| PubChem CID | 10609474 |
| IUPAC Name | 6-chloro-5-(2-chloroethyl)-1,3-dihydroindol-2-one |
| SMILES | C1C2=CC(=C(C=C2NC1=O)Cl)CCCl |
Chemical Identifiers
| 118289-55-7 | |
| 230.088 | |
| ZTQQXEPZEYIVDK-UHFFFAOYSA-N | |
| 10609474 | |
| C1C2=CC(=C(C=C2NC1=O)Cl)CCCl |
| C10H9Cl2NO | |
| MFCD03411598 | |
| 6-Chloro-5-(2-chloroethyl)-2-indolinone | |
| 6-chloro-5-(2-chloroethyl)-1,3-dihydroindol-2-one |
Specifications
| 118289-55-7 | |
| C10H9Cl2NO | |
| 1 g | |
| 6-Chloro-5-(2-chloroethyl)-2-indolinone | |
| C1C2=CC(=C(C=C2NC1=O)Cl)CCCl | |
| 230.088 | |
| 230.09 | |
| Crystalline Powder |
| 225°C | |
| MFCD03411598 | |
| 3077 | |
| ZTQQXEPZEYIVDK-UHFFFAOYSA-N | |
| 6-chloro-5-(2-chloroethyl)-1,3-dihydroindol-2-one | |
| 10609474 | |
| ≥98.0% (GC) | |
| 6-Chloro-5-(2-chloroethyl)oxindole |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator