missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-5-(2-chloroethyl)oxindole 98.0+%, TCI America™

Catalog No. C26511G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26511G Supplier TCI America Supplier No. C26511G

Chemical Identifiers

CAS 118289-55-7
Molecular Formula C10H9Cl2NO
Molecular Weight (g/mol) 230.088
MDL Number MFCD03411598
InChI Key ZTQQXEPZEYIVDK-UHFFFAOYSA-N
Synonym 6-Chloro-5-(2-chloroethyl)-2-indolinone
PubChem CID 10609474
IUPAC Name 6-chloro-5-(2-chloroethyl)-1,3-dihydroindol-2-one
SMILES C1C2=CC(=C(C=C2NC1=O)Cl)CCCl

Specifications

Melting Point 225°C
Quantity 1 g
UN Number 3077
Formula Weight 230.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-5-(2-chloroethyl)oxindole
TSCA No
Recommended Storage Refrigerator