missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-4-chromanone 98.0+%, TCI America™
$150.98 - $452.97
Chemical Identifiers
| CAS | 37674-72-9 |
|---|---|
| Molecular Formula | C9H7ClO2 |
| Molecular Weight (g/mol) | 182.603 |
| InChI Key | LLTDYHFVIVSQPJ-UHFFFAOYSA-N |
| Synonym | 6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5 |
| PubChem CID | 95273 |
| IUPAC Name | 6-chloro-2,3-dihydrochromen-4-one |
| SMILES | C1COC2=C(C1=O)C=C(C=C2)Cl |
Chemical Identifiers
| 37674-72-9 | |
| 182.603 | |
| 6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5 | |
| 6-chloro-2,3-dihydrochromen-4-one |
| C9H7ClO2 | |
| LLTDYHFVIVSQPJ-UHFFFAOYSA-N | |
| 95273 | |
| C1COC2=C(C1=O)C=C(C=C2)Cl |
Specifications
| 37674-72-9 | |
| C9H7ClO2 | |
| 6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5 | |
| C1COC2=C(C1=O)C=C(C=C2)Cl | |
| 182.603 | |
| 182.60 | |
| Crystalline Powder |
| 101°C | |
| 5 g | |
| LLTDYHFVIVSQPJ-UHFFFAOYSA-N | |
| 6-chloro-2,3-dihydrochromen-4-one | |
| 95273 | |
| ≥98.0% (GC) | |
| 6-Chloro-4-chromanone |
Safety and Handling
TSCA : No