missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-3-pyridinemethanol 98.0+%, TCI America™
$121.28 - $365.79
Chemical Identifiers
| CAS | 21543-49-7 |
|---|---|
| Molecular Formula | C6H6ClNO |
| Molecular Weight (g/mol) | 143.57 |
| InChI Key | GOXYBEXWMJZLJB-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-hydroxymethylpyridine, 6-chloropyridin-3-yl methanol, 2-chloro-5-hydroxymethyl pyridine, 2-chloro-5-pyridinemethanol, 6-chloropyridine-3-methanol, 6-chloro-3-pyridinemethanol, 6-chloro-3-pyridinyl methanol, 3-pyridinemethanol, 6-chloro, 6-chloro-3-pyridyl methanol, 6-chloro-3-pyridyl methan-1-ol |
| PubChem CID | 177161 |
| IUPAC Name | (6-chloropyridin-3-yl)methanol |
| SMILES | C1=CC(=NC=C1CO)Cl |
Chemical Identifiers
| 21543-49-7 | |
| 143.57 | |
| 2-chloro-5-hydroxymethylpyridine, 6-chloropyridin-3-yl methanol, 2-chloro-5-hydroxymethyl pyridine, 2-chloro-5-pyridinemethanol, 6-chloropyridine-3-methanol, 6-chloro-3-pyridinemethanol, 6-chloro-3-pyridinyl methanol, 3-pyridinemethanol, 6-chloro, 6-chloro-3-pyridyl methanol, 6-chloro-3-pyridyl methan-1-ol | |
| (6-chloropyridin-3-yl)methanol |
| C6H6ClNO | |
| GOXYBEXWMJZLJB-UHFFFAOYSA-N | |
| 177161 | |
| C1=CC(=NC=C1CO)Cl |
Specifications
| 21543-49-7 | |
| C6H6ClNO | |
| 2-chloro-5-hydroxymethylpyridine, 6-chloropyridin-3-yl methanol, 2-chloro-5-hydroxymethyl pyridine, 2-chloro-5-pyridinemethanol, 6-chloropyridine-3-methanol, 6-chloro-3-pyridinemethanol, 6-chloro-3-pyridinyl methanol, 3-pyridinemethanol, 6-chloro, 6-chloro-3-pyridyl methanol, 6-chloro-3-pyridyl methan-1-ol | |
| C1=CC(=NC=C1CO)Cl | |
| 143.57 | |
| 143.57 | |
| Crystalline Powder |
| 54°C | |
| 5 g | |
| GOXYBEXWMJZLJB-UHFFFAOYSA-N | |
| (6-chloropyridin-3-yl)methanol | |
| 177161 | |
| ≥98.0% (GC) | |
| 6-Chloro-3-pyridinemethanol |
Safety and Handling
TSCA : No