missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-pyridinecarboxaldehyde 96.0+%, TCI America™

Catalog No. C25191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25191G 1 g
C25195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25191G Supplier TCI America Supplier No. C25191G

Chemical Identifiers

CAS 23100-12-1
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.554
MDL Number MFCD03095223
InChI Key AFWWKZCPPRPDQK-UHFFFAOYSA-N
Synonym 6-chloronicotinaldehyde, 2-chloropyridine-5-carbaldehyde, 2-chloropyridine-5-carboxaldehyde, 6-chloropyridine-3-carboxaldehyde, 2-chloro-5-formylpyridine, 6-chloro-pyridine-3-carbaldehyde, 6-chloro-3-pyridinecarboxaldehyde, 2-chloro-5-pyridine carbaldehyde, 6-chloro-pyridin-3-carbaldehyde, 2-chloro-5-pyridinecarboxaldehyde
PubChem CID 2764053
IUPAC Name 6-chloropyridine-3-carbaldehyde
SMILES C1=CC(=NC=C1C=O)Cl

Specifications

Melting Point 79°C
Color Yellow
Quantity 1 g
Formula Weight 141.55
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-3-pyridinecarboxaldehyde