missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-pyridineacetic acid, 97%

Catalog No. AAH2796618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2796618 50 g
AAH2796609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2796618 Supplier Thermo Scientific Chemicals Supplier No. H2796618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39891-13-9 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39891-13-9
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD01863172
InChI Key XWVPSJPQWOVRHJ-UHFFFAOYSA-N
Synonym 2-6-chloropyridin-3-yl acetic acid, 2-chloropyridine-5-acetic acid, 6-chloro-pyridin-3-yl-acetic acid, 6-chloro-3-pyridineacetic acid, 6-chloropyridin-3-yl acetic acid, 2-6-chloro-3-pyridyl acetic acid, 3-pyridineacetic acid, 6-chloro, 2-chloropyridyl-5-acetic acid, 2-2-chloropyridin-5-yl acetic acid, 6-chloropyridine-3-acetic acid
PubChem CID 14071571
IUPAC Name 2-(6-chloropyridin-3-yl)acetic acid
SMILES C1=CC(=NC=C1CC(=O)O)Cl

Specifications

Melting Point 172°C to 176°C
Quantity 50 g
Formula Weight 171.58
Percent Purity 97%
Chemical Name or Material 6-Chloro-3-pyridineacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only