missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-methyluracil 98.0+%, TCI America™

Catalog No. C23005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23005G 5 g
C230025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23005G Supplier TCI America Supplier No. C23005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4318-56-3
Molecular Formula C5H5ClN2O2
Molecular Weight (g/mol) 160.56
MDL Number MFCD01074837,MFCD00829289
InChI Key SGLXGFAZAARYJY-UHFFFAOYSA-N
PubChem CID 96141
IUPAC Name 6-chloro-3-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CN1C(=O)NC(Cl)=CC1=O

Specifications

Melting Point 278°C
Color White-Yellow
Quantity 5 g
Formula Weight 160.56
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-3-methyluracil