missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-3-indolyl beta-D-Galactopyranoside 98.0+%, TCI America™
$48.46 - $136.08
Chemical Identifiers
| CAS | 138182-21-5 |
|---|---|
| Molecular Formula | C14H16ClNO6 |
| Molecular Weight (g/mol) | 329.73 |
| MDL Number | MFCD00467206,MFCD01310928 |
| InChI Key | OQWBAXBVBGNSPW-MBJXGIAVSA-N |
| Synonym | salmon-gal, rose-gal, 6-chloro-3-indolyl beta-d-galactopyranoside |
| PubChem CID | 44630036 |
| IUPAC Name | (2S,3R,4S,5R,6R)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
| SMILES | OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C237120MG
|
TCI America
C237120MG |
20 mg |
Each for $48.46
|
|
|||||
|
C2371100MG
|
TCI America
C2371100MG |
100 mg |
Each for $136.08
|
|
|||||
Chemical Identifiers
| 138182-21-5 | |
| 329.73 | |
| OQWBAXBVBGNSPW-MBJXGIAVSA-N | |
| 44630036 | |
| OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O |
| C14H16ClNO6 | |
| MFCD00467206,MFCD01310928 | |
| salmon-gal, rose-gal, 6-chloro-3-indolyl beta-d-galactopyranoside | |
| (2S,3R,4S,5R,6R)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
Specifications
| 138182-21-5 | |
| C14H16ClNO6 | |
| 20 mg | |
| OQWBAXBVBGNSPW-MBJXGIAVSA-N | |
| (2S,3R,4S,5R,6R)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 44630036 | |
| ≥98.0% (HPLC) | |
| 6-Chloro-3-indolyl beta-D-Galactopyranoside [for Biochemical Research] |
| White-Yellow | |
| MFCD00467206,MFCD01310928 | |
| salmon-gal, rose-gal, 6-chloro-3-indolyl beta-d-galactopyranoside | |
| OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O | |
| 329.73 | |
| 329.73 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer