missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-indolyl beta-D-Galactopyranoside 98.0+%, TCI America™

Catalog No. C237120MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C237120MG 20 mg
C2371100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C237120MG Supplier TCI America Supplier No. C237120MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138182-21-5
Molecular Formula C14H16ClNO6
Molecular Weight (g/mol) 329.73
MDL Number MFCD00467206,MFCD01310928
InChI Key OQWBAXBVBGNSPW-MBJXGIAVSA-N
Synonym salmon-gal, rose-gal, 6-chloro-3-indolyl beta-d-galactopyranoside
PubChem CID 44630036
IUPAC Name (2S,3R,4S,5R,6R)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Color White-Yellow
Quantity 20 mg
Formula Weight 329.73
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-3-indolyl beta-D-Galactopyranoside [for Biochemical Research]
TSCA No
Recommended Storage Freezer