missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-3-indolyl-beta-D-galactopyranoside, 98%

Catalog No. AAB2113177
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2113177 0.1 g
AAB2113102 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2113177 Supplier Thermo Scientific Chemicals Supplier No. B2113177
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 138182-21-5 | C14H16ClNO6 | 329.73 g/mol

Used as a pharmaceutical intermediate to reduce toxicity and as an enzyme substrate in diagnostic reagents. Salmon-gal is used in conjunction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colometric assay to detect recombinants (white) from the non-recombinants (SALMON). In conjunction with X-glu, salmon-gal is useful for simultaneous detection of GUS and Lac activities on the same plate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a pharmaceutical intermediate to reduce toxicity and as an enzyme substrate in diagnostic reagents. Salmon-gal is used in conjunction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colometric assay to detect recombinants (white) from the non-recombinants (SALMON). In conjunction with X-glu, salmon-gal is useful for simultaneous detection of GUS and Lac activities on the same plate.

Solubility
Insoluble in water. Soluble in DMSO and DMFO, methanol.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 138182-21-5
Molecular Formula C14H16ClNO6
Molecular Weight (g/mol) 329.73
MDL Number MFCD00467206,MFCD01310928
InChI Key OQWBAXBVBGNSPW-MBJXGIAVSA-N
Synonym salmon-gal, rose-gal, 6-chloro-3-indolyl beta-d-galactopyranoside
PubChem CID 44630036
IUPAC Name (2S,3S,4S,5R,6S)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Melting Point 188°C to 190°C
Quantity 0.1 g
Sensitivity Moisture sensitive
Solubility Information Insoluble in water. Soluble in DMSO and DMFO,methanol.
Optical Rotation −47° (c=1 in Methanol)
Formula Weight 329.74
Percent Purity 98%
Chemical Name or Material 6-Chloro-3-indolyl-beta-D-galactopyranoside
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only