missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. C30171G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30171G 1 g
C30175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30171G Supplier TCI America Supplier No. C30171G

Chemical Identifiers

CAS 54087-03-5
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.554
MDL Number MFCD09832941
InChI Key XTRLIKXVRGWTKW-UHFFFAOYSA-N
Synonym 6-chloropicolinaldehyde, 6-chloro-2-pyridinecarbaldehyde, 6-chloro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-chloro, 2-chloro-6-formylpyridine, 6-chloro-2-formylpyridine, 2-chloropyridine-6-carboxaldehyde, 6-chloro-2-pyridinecarboxaldehyde, pubchem13323, chloropyridinecarbaldehyde
PubChem CID 10796848
IUPAC Name 6-chloropyridine-2-carbaldehyde
SMILES C1=CC(=NC(=C1)Cl)C=O

Specifications

Melting Point 71°C
Color White
Quantity 1 g
Formula Weight 141.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2-pyridinecarboxaldehyde