missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-2-iodo-3-hydroxypyridine 98.0+%, TCI America™
$150.16 - $444.72
Chemical Identifiers
| CAS | 188057-26-3 |
|---|---|
| Molecular Formula | C5H3ClINO |
| Molecular Weight (g/mol) | 255.439 |
| MDL Number | MFCD07636749 |
| InChI Key | FWIMPBYPMQSSCD-UHFFFAOYSA-N |
| PubChem CID | 23433158 |
| IUPAC Name | 6-chloro-2-iodopyridin-3-ol |
| SMILES | C1=CC(=NC(=C1O)I)Cl |
Chemical Identifiers
| 188057-26-3 | |
| 255.439 | |
| FWIMPBYPMQSSCD-UHFFFAOYSA-N | |
| 6-chloro-2-iodopyridin-3-ol |
| C5H3ClINO | |
| MFCD07636749 | |
| 23433158 | |
| C1=CC(=NC(=C1O)I)Cl |
Specifications
| 188057-26-3 | |
| Yellow | |
| MFCD07636749 | |
| 6-chloro-2-iodo-3-hydroxypyridine, 6-chloro-2-iodo-3-pyridinol, 6-chloro-2-iodo-pyridin-3-ol, 3-pyridinol, 6-chloro-2-iodo, 2-iodo-6-chloropyridine-3-ol, 3-pyridinol,6-chloro-2-iodo | |
| C1=CC(=NC(=C1O)I)Cl | |
| 255.439 | |
| 255.44 | |
| Crystalline Powder |
| 193°C | |
| C5H3ClINO | |
| 1 g | |
| FWIMPBYPMQSSCD-UHFFFAOYSA-N | |
| 6-chloro-2-iodopyridin-3-ol | |
| 23433158 | |
| ≥98.0% (GC,T) | |
| 6-Chloro-2-iodo-3-hydroxypyridine |
Safety and Handling
TSCA : No