missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-iodo-3-hydroxypyridine, 98%

Catalog No. p-7031884
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3312703 1 g
AAH3312709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3312703 Supplier Thermo Scientific Chemicals Supplier No. H3312703

CAS: 188057-26-3 | C5H3ClINO | 255.439 g/mol

2,2'-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.

Solubility
Slightly Soluble (1.3 g/L) (25°C).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Chemical Identifiers

CAS 188057-26-3
Molecular Formula C5H3ClINO
Molecular Weight (g/mol) 255.439
MDL Number MFCD07636749
InChI Key FWIMPBYPMQSSCD-UHFFFAOYSA-N
Synonym 6-chloro-2-iodo-3-hydroxypyridine, 6-chloro-2-iodo-3-pyridinol, 6-chloro-2-iodo-pyridin-3-ol, 3-pyridinol, 6-chloro-2-iodo, 2-iodo-6-chloropyridine-3-ol, 3-pyridinol,6-chloro-2-iodo
PubChem CID 23433158
IUPAC Name 6-chloro-2-iodopyridin-3-ol
SMILES C1=CC(=NC(=C1O)I)Cl

Specifications

Melting Point 191°C to 193°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Slightly Soluble (1.3g/L) (25°C).
Formula Weight 255.44
Percent Purity 98%
Chemical Name or Material 6-Chloro-2-iodo-3-hydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only