missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-hexanone 97.0+%, TCI America™

Catalog No. C133325ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C133325ML 25 mL
C1333250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C133325ML Supplier TCI America Supplier No. C133325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10226-30-9
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00191625
InChI Key CMDIDTNMHQUVPE-UHFFFAOYSA-N
Synonym 6-chloro-2-hexanone, 1-chloro-5-hexanone, 2-hexanone, 6-chloro, 1-chlorohexan-5-one, 1-chlorohexane-5-one, 4-chlorobutyl methyl ketone, chlorohexanone, 1-chloro-hexan-5-one, 6-chloro-hexan-2-one, 6-chloranylhexan-2-one
PubChem CID 82468
IUPAC Name 6-chlorohexan-2-one
SMILES CC(=O)CCCCCl

Specifications

Color Yellow
Boiling Point 79°C
Quantity 25 mL
Formula Weight 134.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Chloro-2-hexanone
TSCA No
Recommended Storage Freezer