missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-hexanone, 98%

Catalog No. p-7035736
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5346206 5 g
AAH5346214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5346206 Supplier Thermo Scientific Chemicals Supplier No. H5346206
Only null left

CAS: 10226-30-9 | C6H11ClO | 134.603 g/mol

6-Chloro-2-hexanone in the synthesis of the keto analogs began with 6-chloro-2-hexanone. Substituted 7-(oxoalkyl)theophyllines were synthesized by alkylation of the respective theophyllines with 6-chloro-2-hexanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-2-hexanone in the synthesis of the keto analogs began with 6-chloro-2-hexanone. Substituted 7-(oxoalkyl)theophyllines were synthesized by alkylation of the respective theophyllines with 6-chloro-2-hexanone.

Solubility
Insoluble in water, soluble in chloroform (chloroform, carbon), n-heptane, ethanol, butanol, etc.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 10226-30-9
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00191625
InChI Key CMDIDTNMHQUVPE-UHFFFAOYSA-N
Synonym 6-chloro-2-hexanone, 1-chloro-5-hexanone, 2-hexanone, 6-chloro, 1-chlorohexan-5-one, 1-chlorohexane-5-one, 4-chlorobutyl methyl ketone, chlorohexanone, 1-chloro-hexan-5-one, 6-chloro-hexan-2-one, 6-chloranylhexan-2-one
PubChem CID 82468
IUPAC Name 6-chlorohexan-2-one
SMILES CC(=O)CCCCCl

Specifications

Density 1.02
Boiling Point 86°C (16 mmHg)
Flash Point 91°C (196°F)
Refractive Index 1.4435
Quantity 5 g
Solubility Information Insoluble in water,soluble in chloroform (chloroform,carbon),n-heptane,ethanol,butanol,etc.
Formula Weight 134.61
Percent Purity 98%
Chemical Name or Material 6-Chloro-2-hexanone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-546-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only