missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-fluoropurine, 97%, Thermo Scientific Chemicals

Catalog No. p-7028485
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25825MC 100 mg
AAH25825ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25825MC Supplier Thermo Scientific Chemicals Supplier No. H25825MC
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1651-29-2
Molecular Formula C5H2ClFN4
Molecular Weight (g/mol) 172.55
MDL Number MFCD02183557
InChI Key UNRIYCIDCQDGQE-UHFFFAOYSA-N
Synonym 6-chloro-2-fluoropurine, 6-chloro-2-fluoro-9h-purine, 2-fluoro-6-chloropurine, 6-chloro-2-fluoro-purine, cfp, 1jdj, pubchem9261, acmc-1c2uh, 2-fluoro,6-chloro-purine, 2-fluoro-6-chloro-purine
PubChem CID 5287914
IUPAC Name 6-chloro-2-fluoro-7H-purine
SMILES FC1=NC(Cl)=C2NC=NC2=N1

Specifications

Melting Point 151°C to 153°C
Quantity 100 mg
Formula Weight 172.55
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoropurine
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only