Learn More
6-Chloro-2,4-dimethoxypyrimidine, 98+%
CAS: 6320-15-6 | C6H7ClN2O2 | 174.58 g/mol
$100.26 - $335.41
Chemical Identifiers
| CAS | 6320-15-6 |
|---|---|
| Molecular Formula | C6H7ClN2O2 |
| Molecular Weight (g/mol) | 174.58 |
| MDL Number | MFCD00006068 |
| InChI Key | JHNRTJRDRWKAIW-UHFFFAOYSA-N |
| Synonym | 6-chloro-2,4-dimethoxypyrimidine, 2,4-dimethoxy-6-chloropyrimidine, pyrimidine, 4-chloro-2,6-dimethoxy, 4-chloro-2,6-dimethoxy-pyrimidine, pubchem16350, acmc-209ndu, 4-chlor-2,6-dimethoxypyrimidin, 4-chloro-2,6-dimethoxypyrimidin, 2,6-dimethoxy-4-chloropyrimidine, 6-chloro-2,4-dimethixypyrimidine |
| PubChem CID | 80600 |
| IUPAC Name | 4-chloro-2,6-dimethoxypyrimidine |
| SMILES | COC1=CC(Cl)=NC(OC)=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2169606
|
Thermo Scientific Chemicals
B2169606 |
5 g |
Each for $100.26
|
|
|||||
|
AAB2169614
|
Thermo Scientific Chemicals
B2169614 |
25 g |
Each for $335.41
|
|
|||||
Description
It is used in the synthesis of silyl and stannyl pyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the synthesis of silyl and stannyl pyrimidines.
Solubility
Soluble in chloroform, ethyl acetate and methanol.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 6320-15-6 | |
| 174.58 | |
| JHNRTJRDRWKAIW-UHFFFAOYSA-N | |
| 80600 | |
| COC1=CC(Cl)=NC(OC)=N1 |
| C6H7ClN2O2 | |
| MFCD00006068 | |
| 6-chloro-2,4-dimethoxypyrimidine, 2,4-dimethoxy-6-chloropyrimidine, pyrimidine, 4-chloro-2,6-dimethoxy, 4-chloro-2,6-dimethoxy-pyrimidine, pubchem16350, acmc-209ndu, 4-chlor-2,6-dimethoxypyrimidin, 4-chloro-2,6-dimethoxypyrimidin, 2,6-dimethoxy-4-chloropyrimidine, 6-chloro-2,4-dimethixypyrimidine | |
| 4-chloro-2,6-dimethoxypyrimidine |
Specifications
| 6320-15-6 | |
| C6H7ClN2O2 | |
| 5 g | |
| Hygroscopic | |
| Soluble in chloroform,ethyl acetate and methanol. | |
| COC1=CC(Cl)=NC(OC)=N1 | |
| 174.58 | |
| 174.59 | |
| 6-Chloro-2,4-dimethoxypyrimidine |
| 71°C to 73°C | |
| MFCD00006068 | |
| 137581 | |
| 6-chloro-2,4-dimethoxypyrimidine, 2,4-dimethoxy-6-chloropyrimidine, pyrimidine, 4-chloro-2,6-dimethoxy, 4-chloro-2,6-dimethoxy-pyrimidine, pubchem16350, acmc-209ndu, 4-chlor-2,6-dimethoxypyrimidin, 4-chloro-2,6-dimethoxypyrimidin, 2,6-dimethoxy-4-chloropyrimidine, 6-chloro-2,4-dimethixypyrimidine | |
| JHNRTJRDRWKAIW-UHFFFAOYSA-N | |
| 4-chloro-2,6-dimethoxypyrimidine | |
| 80600 | |
| ≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-669-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only