missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2,4-diaminopyrimidine 98.0+%, TCI America™

Catalog No. C081125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C081125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C081125G Supplier TCI America Supplier No. C081125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 156-83-2
Molecular Formula C4H5ClN4
Molecular Weight (g/mol) 144.562
MDL Number MFCD00006097
InChI Key QJIUMVUZDYPQRT-UHFFFAOYSA-N
Synonym 4-chloro-2,6-diaminopyrimidine, 2,4-diamino-6-chloropyrimidine, 2,6-diamino-4-chloropyrimidine, 6-chloro-2,4-diaminopyrimidine, 6-chloro-pyrimidine-2,4-diamine, 2,4-pyrimidinediamine, 6-chloro, 4-chloro-2,6-diamino pyrimidine, 2-amino-6-chloro-4-pyrimidinylamine, caap, minoxidil impurity b
PubChem CID 67432
IUPAC Name 6-chloropyrimidine-2,4-diamine
SMILES C1=C(N=C(N=C1Cl)N)N

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 25 g
Formula Weight 144.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2,4-diaminopyrimidine
EINECSNumber (5)-5972
TSCA No