missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1-hexene, 97%

Catalog No. p-7035673
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5339606 5 g
AAH5339614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5339606 Supplier Thermo Scientific Chemicals Supplier No. H5339606
Only null left

CAS: 928-89-2 | C6H11Cl | 118.604 g/mol

6-Chloro-1-hexene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chloro-1-hexene is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 928-89-2
Molecular Formula C6H11Cl
Molecular Weight (g/mol) 118.604
MDL Number MFCD00039391
InChI Key BLMIXWDJHNJWDT-UHFFFAOYSA-N
Synonym 6-chloro-1-hexene, 1-hexene, 6-chloro, 5-hexenyl chloride, 6-chlorohexene, chloro-1-hex, 6-chloro-1-hexen, 1-chloro-5-hexene, 1-chlorohex-5-ene, acmc-20alz4, 5-hexen-1-yl chloride
PubChem CID 70233
IUPAC Name 6-chlorohex-1-ene
SMILES C=CCCCCCl

Specifications

Density 0.896
Boiling Point 135°C to 136°C
Flash Point 29°C (84°F)
Refractive Index 1.438
Linear Formula Cl(CH2)4CH=CH2
Quantity 5 g
UN Number UN1993
Formula Weight 118.61
Percent Purity 97%
Chemical Name or Material 6-Chloro-1-hexene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 213-186-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only