missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1-hexanol 96.0+%, TCI America™

Catalog No. C080125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C080125G 25 g
C0801500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C080125G Supplier TCI America Supplier No. C080125G

Chemical Identifiers

CAS 2009-83-8
Molecular Formula C6H13ClO
Molecular Weight (g/mol) 136.619
MDL Number MFCD00002984
InChI Key JNTPTNNCGDAGEJ-UHFFFAOYSA-N
Synonym 6-chloro-1-hexanol, 6-chlorohexanol, 1-chloro-6-hydroxyhexane, 1-hexanol, 6-chloro, hexamethylene chlorohydrin, 1-chloro-6-hexanol, omega-chlorohexanol, unii-t2hit6n5uf, 6-chloro-hexanol, t2hit6n5uf
PubChem CID 74828
IUPAC Name 6-chlorohexan-1-ol
SMILES C(CCCCl)CCO

Specifications

Color Yellow
Boiling Point 117°C
Quantity 25 g
Formula Weight 136.62
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 6-Chloro-1-hexanol
EINECSNumber (2)-3811
RTECSNumber MO9160000
TSCA Yes