Learn More
6-Chloro-1-hexanol, 97%
CAS: 2009-83-8 | C6H13ClO | 136.619 g/mol
Supplier: Thermo Scientific Chemicals A1458222
Description
6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Chloro-1-hexanol is used in the preparation of its corresponding bromide by reaction with sodium bromide using N,N-dimethylformamide and dibromomethane (2:1) as a solvent. It is utilized in the selective elimination of 6-chloro-1-hexene from esters of 6-chloro-1-hexanol.
Solubility
Soluble in dimethylformamide.
Notes
Incompatible with strong oxidizing agents, strong bases, powdered metal salts, strong reducing agents, acid anhydrides and acid chlorides.
Specifications
| 6-Chloro-1-hexanol | |
| 1.027 | |
| 98°C (208°F) | |
| 1.456 | |
| 100 g | |
| 6-chloro-1-hexanol, 6-chlorohexanol, 1-chloro-6-hydroxyhexane, 1-hexanol, 6-chloro, hexamethylene chlorohydrin, 1-chloro-6-hexanol, omega-chlorohexanol, unii-t2hit6n5uf, 6-chloro-hexanol, t2hit6n5uf | |
| JNTPTNNCGDAGEJ-UHFFFAOYSA-N | |
| 6-chlorohexan-1-ol | |
| 74828 | |
| 97% |
| 2009-83-8 | |
| 109°C to 111°C (14 mmHg) | |
| C6H13ClO | |
| MFCD00002984 | |
| 1697307 | |
| Soluble in dimethylformamide. | |
| C(CCCCl)CCO | |
| 136.619 | |
| 136.62 |
Chemical Identifiers
| 2009-83-8 | |
| 136.619 | |
| JNTPTNNCGDAGEJ-UHFFFAOYSA-N | |
| 74828 | |
| C(CCCCl)CCO |
| C6H13ClO | |
| MFCD00002984 | |
| 6-chloro-1-hexanol, 6-chlorohexanol, 1-chloro-6-hydroxyhexane, 1-hexanol, 6-chloro, hexamethylene chlorohydrin, 1-chloro-6-hexanol, omega-chlorohexanol, unii-t2hit6n5uf, 6-chloro-hexanol, t2hit6n5uf | |
| 6-chlorohexan-1-ol |
Safety and Handling
GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 217-925-2
RTECSNumber : MO9160000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only