missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1,3-dimethyluracil, 97%

Catalog No. AAA1275506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1275506 5 g
AAA1275503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1275506 Supplier Thermo Scientific Chemicals Supplier No. A1275506
Only null left

CAS: 6972-27-6 | C6H7ClN2O2 | 174.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6972-27-6
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.58
MDL Number MFCD00038066
InChI Key VATQPUHLFQHDBD-UHFFFAOYSA-N
Synonym 6-chloro-1,3-dimethyluracil, 6-chloro-1,3-dimethylpyrimidine-2,4 1h,3h-dione, 1,3-dimethyl-6-chlorouracil, 6-chloro-1,3-dimethyl-2,4 1h,3h-pyrimidinedione, 2,4 1h,3h-pyrimidinedione, 6-chloro-1,3-dimethyl, 6-chloro-2,4-dihydroxy-1,3-dimethylpyrimidine, 6-chloro-1,3-dimethyl-2,4-1h,3h-pyrimidinedione, 6-chloro-1,3-dimethyl-1,3-dihydropyrimidine-2,4-dione, acmc-209oan, 1,3-dimethyl-4-chlorouracil
PubChem CID 81442
IUPAC Name 6-chloro-1,3-dimethylpyrimidine-2,4-dione
SMILES CN1C(Cl)=CC(=O)N(C)C1=O

Specifications

Melting Point 109°C to 111°C
Quantity 5 g
Beilstein 144393
Formula Weight 174.59
Percent Purity 97%
Chemical Name or Material 6-Chloro-1,3-dimethyluracil
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-205-2
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only