missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoquinazoline-2,4(1H,3H)-dione, 97%

Catalog No. AAH6354703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6354703 1 g
AAH6354706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6354703 Supplier Thermo Scientific Chemicals Supplier No. H6354703

CAS: 88145-89-5 | C8H5BrN2O2 | 241.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88145-89-5
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.044
MDL Number MFCD00462868
InChI Key JZDVFUAHGLJVQG-UHFFFAOYSA-N
Synonym 6-bromoquinazoline-2,4 1h,3h-dione, 6-bromo-2,4 1h,3h-quinazolinedione, 6-bromoquinazoline-2,4-dione, 6-bromoquinazoline-2,4-diol, 2,4 1h,3h-quinazolinedione, 6-bromo, 6-bromo-1,3-dihydroquinazoline-2,4-dione, 6-bromo-1,2,3,4-tetrahydroquinazoline-2,4-dione, bromoquinazolinedione, pubchem20961, acmc-209qre
PubChem CID 617686
IUPAC Name 6-bromo-1H-quinazoline-2,4-dione
SMILES C1=CC2=C(C=C1Br)C(=O)NC(=O)N2

Specifications

Color White
Quantity 1 g
Formula Weight 241.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Bromoquinazoline-2,4(1H,3H)-dione
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only