Learn More
6-Bromopyridine-3-boronic acid, 95%
CAS: 223463-14-7 | C5H5BBrNO2 | 201.81 g/mol
$99.03 - $1,076.84
Chemical Identifiers
| CAS | 223463-14-7 |
|---|---|
| Molecular Formula | C5H5BBrNO2 |
| Molecular Weight (g/mol) | 201.81 |
| MDL Number | MFCD03411558 |
| InChI Key | BCYWDUVHAPHGIP-UHFFFAOYSA-N |
| Synonym | 2-bromopyridine-5-boronic acid, 6-bromopyridin-3-yl boronic acid, 6-bromopyridine-3-boronic acid, 2-bromo-5-pyridineboronic acid, 6-bromopyridin-3-lboronic acid, 2-bromo-5-pyridylboronic acid, 2-bromopyridyl-5-boronic acid, 6-bromo-3-pyridylboronic acid, 6-bromopyridin-3-yl-3-boronic acid, 6-bromo-3-pyridinylboronic acid |
| PubChem CID | 2762706 |
| IUPAC Name | (6-bromopyridin-3-yl)boronic acid |
| SMILES | OB(O)C1=CN=C(Br)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL20085MD
|
Thermo Scientific Chemicals
L20085MD |
250 mg |
N/A
|
|
|||||
|
AAL2008503
|
Thermo Scientific Chemicals
L2008503 |
1 g |
N/A
|
|
|||||
|
AAL2008506
|
Thermo Scientific Chemicals
L2008506 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 223463-14-7 | |
| 201.81 | |
| BCYWDUVHAPHGIP-UHFFFAOYSA-N | |
| 2762706 | |
| OB(O)C1=CN=C(Br)C=C1 |
| C5H5BBrNO2 | |
| MFCD03411558 | |
| 2-bromopyridine-5-boronic acid, 6-bromopyridin-3-yl boronic acid, 6-bromopyridine-3-boronic acid, 2-bromo-5-pyridineboronic acid, 6-bromopyridin-3-lboronic acid, 2-bromo-5-pyridylboronic acid, 2-bromopyridyl-5-boronic acid, 6-bromo-3-pyridylboronic acid, 6-bromopyridin-3-yl-3-boronic acid, 6-bromo-3-pyridinylboronic acid | |
| (6-bromopyridin-3-yl)boronic acid |
Specifications
| 223463-14-7 | |
| C5H5BBrNO2 | |
| 250 mg | |
| 2-bromopyridine-5-boronic acid, 6-bromopyridin-3-yl boronic acid, 6-bromopyridine-3-boronic acid, 2-bromo-5-pyridineboronic acid, 6-bromopyridin-3-lboronic acid, 2-bromo-5-pyridylboronic acid, 2-bromopyridyl-5-boronic acid, 6-bromo-3-pyridylboronic acid, 6-bromopyridin-3-yl-3-boronic acid, 6-bromo-3-pyridinylboronic acid | |
| OB(O)C1=CN=C(Br)C=C1 | |
| 201.81 | |
| 201.82 | |
| 6-Bromopyridine-3-boronic acid |
| ∽165°C (decomposition) | |
| MFCD03411558 | |
| 9119918 | |
| BCYWDUVHAPHGIP-UHFFFAOYSA-N | |
| (6-bromopyridin-3-yl)boronic acid | |
| 2762706 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H312-H315-H319-H332-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only