missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyridine-2-carboxamide, 97%

Catalog No. AAH3181103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3181103 1 g
AAH3181106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3181103 Supplier Thermo Scientific Chemicals Supplier No. H3181103
Only null left

CAS: 25194-52-9 | C6H5BrN2O | 201.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25194-52-9
Molecular Formula C6H5BrN2O
Molecular Weight (g/mol) 201.02
MDL Number MFCD00234154
InChI Key HWRUOHMDYQEWOD-UHFFFAOYSA-N
PubChem CID 21961408
IUPAC Name 6-bromopyridine-2-carboxamide
SMILES NC(=O)C1=NC(Br)=CC=C1

Specifications

Quantity 1 g
Synonym 6-bromopicolinamide, 6-bromopyridine-2-carboxylic acid amide, 6-bromo-2-pyridinecarboxamide, 2-pyridinecarboxamide, 6-bromo, 6-bromo-pyridine-2-carboxylic acid amide, 6-bromo-2-carbamoylpyridine, 2-pyridinecarboxamide,6-bromo, acmc-209gi0, tpc-i169, 6-bromanylpyridine-2-carboxamide
Formula Weight 201.02
Percent Purity 97%
Chemical Name or Material 6-Bromopyridine-2-carboxamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only