missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyridine-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH2724503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2724503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2724503 Supplier Thermo Scientific Chemicals Supplier No. H2724503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 651358-83-7
Molecular Formula C11H15BBrNO2
Molecular Weight (g/mol) 283.96
MDL Number MFCD06796177
InChI Key OAVRLCKBKDMGGW-UHFFFAOYSA-N
Synonym 6-bromopyridine-2-boronic acid pinacol ester, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-6-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromopyridine-6-boronic acid pinacol ester, 2-6-bromo-2-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine, 2-bromo-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-bromo-2-pyridylboronic acid pinacol ester, 6-bromo-2-pyridineboronic acid pinacol ester, 6-bromopyridin-2-ylboronic acid pinacol ester, 6-bromopyridin-2-yl boronic acid pinacol ester
PubChem CID 12177444
IUPAC Name 2-bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=NC(=CC=C2)Br

Specifications

Melting Point 125°C to 127°C
Quantity 1 g
Formula Weight 283.96
Percent Purity 95%
Chemical Name or Material 6-Bromopyridine-2-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only