missing translation for 'onlineSavingsMsg'
Learn More

6-Bromopyrazolo[1,5-a]pyrimidine, 97%

Catalog No. AAH3385506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3385506 5 g
AAH3385503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3385506 Supplier Thermo Scientific Chemicals Supplier No. H3385506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 705263-10-1 | C6H4BrN3 | 198.02 g/mol

6-Bromopyrazolo[1,5-a]pyrimidine is a reactant in the synthesis of LDN-212854, a selective and potent inhibitor of the BMP type I receptor kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopyrazolo[1,5-a]pyrimidine is a reactant in the synthesis of LDN-212854, a selective and potent inhibitor of the BMP type I receptor kinases.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.

Chemical Identifiers

CAS 705263-10-1
Molecular Formula C6H4BrN3
Molecular Weight (g/mol) 198.02
MDL Number MFCD09832894
InChI Key VDHTXLUCUNPVLO-UHFFFAOYSA-N
Synonym 6-bromopyrazolo 1,5-a pyrimidine, 6-bromo-pyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine, 6-bromo, pubchem15248, acmc-209of0
PubChem CID 22236701
IUPAC Name 6-bromopyrazolo[1,5-a]pyrimidine
SMILES BrC1=CN2N=CC=C2N=C1

Specifications

Melting Point 122°C to 124°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 198.02
Percent Purity 97%
Chemical Name or Material 6-Bromopyrazolo[1,5-a]pyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only