missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(Bromomethyl)-3,4-dihydro-2H-1,5-benzodioxepine, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 250 mg |
| Formula Weight | 243.1 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-(Bromomethyl)-3, 4-dihydro-2H-1, 5-benzodioxepine |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS P Statement Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): |