missing translation for 'onlineSavingsMsg'
Learn More

6-(Bromomethyl)-2-pyridinemethanol 98.0+%, TCI America™

Catalog No. p-7119384
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19631G 1 g
B19635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19631G Supplier TCI America Supplier No. B19631G
Only null left

Chemical Identifiers

CAS 40054-01-1
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD01631297
InChI Key JLYYQZROGJLCCS-UHFFFAOYSA-N
Synonym 2-(Bromomethyl)-6-(hydroxymethyl)pyridine
PubChem CID 10878213
IUPAC Name [6-(bromomethyl)pyridin-2-yl]methanol
SMILES C1=CC(=NC(=C1)CBr)CO

Specifications

Melting Point 80°C
Quantity 1 g
Formula Weight 202.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-(Bromomethyl)-2-pyridinemethanol