missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoisoquinoline 98.0+%, TCI America™

Catalog No. b47151g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47151G Supplier TCI America Supplier No. B47151G
Only null left

Chemical Identifiers

CAS 34784-05-9
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.058
MDL Number MFCD04973299
InChI Key ZTEATMVVGQUULZ-UHFFFAOYSA-N
Synonym 6-bromo-isoquinoline, isoquinoline, 6-bromo, 6-bromo isoquinoline, zlchem 769, 6-bromoisoquinoline, pubchem17031, acmc-209ia9, ksc201e8h, monohydrate 1g
PubChem CID 313681
IUPAC Name 6-bromoisoquinoline
SMILES C1=CC2=C(C=CN=C2)C=C1Br

Specifications

Melting Point 46°C
Quantity 1 g
Formula Weight 208.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoisoquinoline