missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoisatin 97.0+%, TCI America™

Catalog No. p-7119666
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24245G 5 g
B242425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24245G Supplier TCI America Supplier No. B24245G
Only null left

Chemical Identifiers

CAS 6326-79-0
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.03
MDL Number MFCD01631138
InChI Key HVPQMLZLINVIHW-UHFFFAOYSA-N
Synonym 6-bromoisatin, 6-bromoindoline-2,3-dione, 6-bromo-isatin, 1h-indole-2,3-dione, 6-bromo, 6-bromoindole-2,3-dione, 6-bromo-2,3-dihydro-1h-indole-2,3-dione, 6-bromo-1h-benzo d azolidine-2,3-dione, 6-bromoindolin-2,3-dione, 6-bromo-2,3-indolinedione
PubChem CID 95716
IUPAC Name 6-bromo-2,3-dihydro-1H-indole-2,3-dione
SMILES BrC1=CC=C2C(NC(=O)C2=O)=C1

Specifications

Melting Point 274°C
Quantity 5 g
Formula Weight 226.03
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoisatin