missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoisatin, 95%

CAS: 6326-79-0 | C8H4BrNO2 | 226.03 g/mol

$527.51 - $527.51

Chemical Identifiers

CAS 6326-79-0
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.03
MDL Number MFCD01631138
InChI Key HVPQMLZLINVIHW-UHFFFAOYSA-N
Synonym 6-bromoisatin, 6-bromoindoline-2,3-dione, 6-bromo-isatin, 1h-indole-2,3-dione, 6-bromo, 6-bromoindole-2,3-dione, 6-bromo-2,3-dihydro-1h-indole-2,3-dione, 6-bromo-1h-benzo d azolidine-2,3-dione, 6-bromoindolin-2,3-dione, 6-bromo-2,3-indolinedione
PubChem CID 95716
SMILES BrC1=CC=C2C(NC(=O)C2=O)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC455510250
SDS View Documents
Thermo Scientific Chemicals
455510250
25 g
Each for $527.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6326-79-0
226.03
HVPQMLZLINVIHW-UHFFFAOYSA-N
95716
C8H4BrNO2
MFCD01631138
6-bromoisatin, 6-bromoindoline-2,3-dione, 6-bromo-isatin, 1h-indole-2,3-dione, 6-bromo, 6-bromoindole-2,3-dione, 6-bromo-2,3-dihydro-1h-indole-2,3-dione, 6-bromo-1h-benzo d azolidine-2,3-dione, 6-bromoindolin-2,3-dione, 6-bromo-2,3-indolinedione
BrC1=CC=C2C(NC(=O)C2=O)=C1

Specifications

6326-79-0
Authentic
C8H4BrNO2
25 g
HVPQMLZLINVIHW-UHFFFAOYSA-N
6-bromo-1H-indole-2,3-dione
95716
95%
274.0°C
94% min. (HPLC)
MFCD01631138
6-bromoisatin, 6-bromoindoline-2,3-dione, 6-bromo-isatin, 1h-indole-2,3-dione, 6-bromo, 6-bromoindole-2,3-dione, 6-bromo-2,3-dihydro-1h-indole-2,3-dione, 6-bromo-1h-benzo d azolidine-2,3-dione, 6-bromoindolin-2,3-dione, 6-bromo-2,3-indolinedione
BrC1=CC=C2C(NC(=O)C2=O)=C1
226.03
226.03
6-Bromoisatin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only