missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoisatin, 95%

Catalog No. AC455510050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455510050 5 g
AC455510250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455510050 Supplier Thermo Scientific Chemicals Supplier No. 455510050

CAS: 6326-79-0 | C8H4BrNO2 | 226.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6326-79-0
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.03
MDL Number MFCD01631138
InChI Key HVPQMLZLINVIHW-UHFFFAOYSA-N
Synonym 6-bromoisatin, 6-bromoindoline-2,3-dione, 6-bromo-isatin, 1h-indole-2,3-dione, 6-bromo, 6-bromoindole-2,3-dione, 6-bromo-2,3-dihydro-1h-indole-2,3-dione, 6-bromo-1h-benzo d azolidine-2,3-dione, 6-bromoindolin-2,3-dione, 6-bromo-2,3-indolinedione
PubChem CID 95716
SMILES BrC1=CC=C2C(NC(=O)C2=O)=C1

Specifications

Melting Point 274.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
IUPAC Name 6-bromo-2,3-dihydro-1H-indole-2,3-dione
Formula Weight 226.03
Percent Purity 95%
Chemical Name or Material 6-Bromoisatin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

RUO – Research Use Only