missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromoindole-3-carboxaldehyde 98.0+%, TCI America™
$137.94 - $467.87
Chemical Identifiers
| CAS | 17826-04-9 |
|---|---|
| Molecular Formula | C9H6BrNO |
| Molecular Weight (g/mol) | 224.06 |
| MDL Number | MFCD00792689 |
| InChI Key | WCCLQCBKBPTODV-UHFFFAOYSA-N |
| Synonym | 6-bromoindole-3-carboxaldehyde, 6-bromoindole-3-carbaldehyde, 6-bromo-3-formylindole, 6-bromo-1h-indole-3-carboxaldehyde, 6-bromindol-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 6-bromo, pubchem7691, acmc-209ee9, ksc536o3d, wcclqcbkbptodv-uhfffaoysa |
| PubChem CID | 2794830 |
| IUPAC Name | 6-bromo-1H-indole-3-carbaldehyde |
| SMILES | BrC1=CC=C2C(NC=C2C=O)=C1 |
Chemical Identifiers
| 17826-04-9 | |
| 224.06 | |
| WCCLQCBKBPTODV-UHFFFAOYSA-N | |
| 2794830 | |
| BrC1=CC=C2C(NC=C2C=O)=C1 |
| C9H6BrNO | |
| MFCD00792689 | |
| 6-bromoindole-3-carboxaldehyde, 6-bromoindole-3-carbaldehyde, 6-bromo-3-formylindole, 6-bromo-1h-indole-3-carboxaldehyde, 6-bromindol-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 6-bromo, pubchem7691, acmc-209ee9, ksc536o3d, wcclqcbkbptodv-uhfffaoysa | |
| 6-bromo-1H-indole-3-carbaldehyde |
Specifications
| 17826-04-9 | |
| C9H6BrNO | |
| 1 g | |
| WCCLQCBKBPTODV-UHFFFAOYSA-N | |
| 6-bromo-1H-indole-3-carbaldehyde | |
| 2794830 | |
| ≥98.0% (GC,N) | |
| 6-Bromoindole-3-carboxaldehyde |
| 203°C | |
| MFCD00792689 | |
| 6-bromoindole-3-carboxaldehyde, 6-bromoindole-3-carbaldehyde, 6-bromo-3-formylindole, 6-bromo-1h-indole-3-carboxaldehyde, 6-bromindol-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 6-bromo, pubchem7691, acmc-209ee9, ksc536o3d, wcclqcbkbptodv-uhfffaoysa | |
| BrC1=CC=C2C(NC=C2C=O)=C1 | |
| 224.06 | |
| 224.06 | |
| Crystalline Powder |
Safety and Handling
TSCA : No