missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoindole-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. B27781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27781G 1 g
B27785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27781G Supplier TCI America Supplier No. B27781G

Chemical Identifiers

CAS 17826-04-9
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.06
MDL Number MFCD00792689
InChI Key WCCLQCBKBPTODV-UHFFFAOYSA-N
Synonym 6-bromoindole-3-carboxaldehyde, 6-bromoindole-3-carbaldehyde, 6-bromo-3-formylindole, 6-bromo-1h-indole-3-carboxaldehyde, 6-bromindol-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 6-bromo, pubchem7691, acmc-209ee9, ksc536o3d, wcclqcbkbptodv-uhfffaoysa
PubChem CID 2794830
IUPAC Name 6-bromo-1H-indole-3-carbaldehyde
SMILES BrC1=CC=C2C(NC=C2C=O)=C1

Specifications

Melting Point 203°C
Quantity 1 g
Formula Weight 224.06
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoindole-3-carboxaldehyde