missing translation for 'onlineSavingsMsg'
Learn More

6-Bromoimidazo[1,2-a]pyridine, 98%

Catalog No. p-7032828
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3445303 1 g
AAH3445306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3445303 Supplier Thermo Scientific Chemicals Supplier No. H3445303
Only null left

CAS: 6188-23-4 | C7H5BrN2 | 197.035 g/mol

It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.

Solubility
slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 6188-23-4
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD06496200
InChI Key FXPMFQUOGYGTAM-UHFFFAOYSA-N
Synonym 6-bromoimidazo 1,2-a pyridine, 6-bromoh-imidazo 1,2-a pyridine, 6-bromo-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 6-bromo, buttpark 154\50-43, pubchem14736, acmc-209mx4, 6-bromo-4-hydroimidazo 1,2-a pyridine
PubChem CID 2795547
IUPAC Name 6-bromoimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=CN2C=C1Br

Specifications

Melting Point 78°C to 80°C
Quantity 1 g
Solubility Information slightly soluble in water.
Formula Weight 197.03
Percent Purity 98%
Chemical Name or Material 6-Bromoimidazo[1,2-a]pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only