missing translation for 'onlineSavingsMsg'
Learn More

6-Bromohexanoic Acid 98.0+%, TCI America™

Catalog No. B1290250G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1290250G 250 g
B129025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1290250G Supplier TCI America Supplier No. B1290250G
Only null left

Chemical Identifiers

CAS 4224-70-8
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00004422
InChI Key NVRVNSHHLPQGCU-UHFFFAOYSA-N
Synonym 6-bromocaproic acid, hexanoic acid, 6-bromo, 6-bromo-hexanoic acid, 6-bromo-n-caproic acid, 6-bromo hexanoic acid, epsilon-bromo caproic acid, 6-bromo hexanoicacid, 6bromohexanoic acid, 6-bromo-hexanicacid, 6-bromo-caproic acid
PubChem CID 20210
ChEBI CHEBI:60700
IUPAC Name 6-bromohexanoic acid
SMILES C(CCC(=O)O)CCBr

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 170°C
Quantity 250 g
UN Number 3261
Formula Weight 195.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromohexanoic Acid
TSCA Yes
Recommended Storage Refrigerator