missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromohexanoic acid, 98+%
Description
6-Bromohexanoic acid is used in the preparation of N-acylsulfonamides, diarylthioethers and 2-lithio-1,3-dithian. It is widely used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Bromohexanoic acid is used in the preparation of N-acylsulfonamides, diarylthioethers and 2-lithio-1,3-dithian. It is widely used as an intermediate in organic synthesis.
Solubility
Soluble in methanol.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with strong oxidizing agents.
6-Bromohexanoic acid is used in the preparation of N-acylsulfonamides, diarylthioethers and 2-lithio-1,3-dithian. It is widely used as an intermediate in organic synthesis.
Solubility
Soluble in methanol.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 32°C to 35°C |
| Boiling Point | 165°C to 170°C (20 mmHg) |
| Flash Point | 95°C (203°F) |
| Odor | Odorless |
| Quantity | 25 g |
| UN Number | UN3261 |
| Beilstein | 1749740 |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 195.06 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 224-176-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only