missing translation for 'onlineSavingsMsg'
Learn More

6-Bromochromone-2-carboxylic Acid 95.0+%, TCI America™

Catalog No. B48021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48021G 1 g
B48025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48021G Supplier TCI America Supplier No. B48021G

Chemical Identifiers

CAS 51484-06-1
Molecular Formula C10H5BrO4
Molecular Weight (g/mol) 269.05
MDL Number MFCD01242619
InChI Key QSBZDBNPXSVVHH-UHFFFAOYSA-N
Synonym 6-bromochromone-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylic acid, 6-bromo-4-oxo-4h-1-benzopyran-2-carboxylic acid, acmc-20alzt, 6-bromo-4-oxo-4h-chromene-2-carboxylicacid, 6-bromochromone-2-carboxylicacid, 6-brornochrornone-2-carboxylic acid
PubChem CID 688867
IUPAC Name 6-bromo-4-oxo-4H-chromene-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C2=CC(Br)=CC=C2O1

Specifications

Melting Point 276°C
Color White-Yellow
Quantity 1 g
Formula Weight 269.05
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromochromone-2-carboxylic Acid