missing translation for 'onlineSavingsMsg'
Learn More

6-Bromochroman, 97%, Thermo Scientific™

Chemical Identifiers

CAS 3875-78-3
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD10698725
InChI Key KPFDABVKWKOIME-UHFFFAOYSA-N
Synonym 6-bromochroman, 6-bromo-chroman, 6-bromochromane, 6-bromo-3,4-dihydro-2h-1-benzopyran, 2h-1-benzopyran,6-bromo-3,4-dihydro, 6-bromanyl-3,4-dihydro-2h-chromene, 2h-1-benzopyran, 6-bromo-3,4-dihydro
PubChem CID 10856814
IUPAC Name 6-bromo-3,4-dihydro-2H-chromene
SMILES C1CC2=C(C=CC(=C2)Br)OC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC32810DA
SDS View Documents
Thermo Scientific Chemicals
CC32810DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC32810DE
SDS View Documents
Thermo Scientific Chemicals
CC32810DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

3875-78-3
213.074
KPFDABVKWKOIME-UHFFFAOYSA-N
10856814
C1CC2=C(C=CC(=C2)Br)OC1
C9H9BrO
MFCD10698725
6-bromochroman, 6-bromo-chroman, 6-bromochromane, 6-bromo-3,4-dihydro-2h-1-benzopyran, 2h-1-benzopyran,6-bromo-3,4-dihydro, 6-bromanyl-3,4-dihydro-2h-chromene, 2h-1-benzopyran, 6-bromo-3,4-dihydro
6-bromo-3,4-dihydro-2H-chromene

Specifications

3875-78-3
100.0
Amber Glass Bottle
MFCD10698725
6-bromochroman, 6-bromo-chroman, 6-bromochromane, 6-bromo-3,4-dihydro-2h-1-benzopyran, 2h-1-benzopyran,6-bromo-3,4-dihydro, 6-bromanyl-3,4-dihydro-2h-chromene, 2h-1-benzopyran, 6-bromo-3,4-dihydro
C1CC2=C(C=CC(=C2)Br)OC1
213.074
213.07
6-Bromochroman
96.5
95%
C9H9BrO
1g
KPFDABVKWKOIME-UHFFFAOYSA-N
6-bromo-3,4-dihydro-2H-chromene
10856814
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'