missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-hydroxyquinoline 98.0+%, TCI America™

$119.36 - $119.36

Chemical Identifiers

CAS 145369-94-4
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD00272435
InChI Key XKLBNOHKHRAXKK-UHFFFAOYSA-N
Synonym 6-bromoquinolin-4-ol, 6-bromo-4-hydroxyquinoline, 6-bromoquinolin-4 1h-one, 6-bromo-quinolin-4-ol, 4-quinolinol, 6-bromo, 6-bromo-1,4-dihydroquinolin-4-one, 6-bromo-4-quinolinol, 4-hydroxy-6-bromoquinoline, 6-bromo-4-hyrdroxyquinoline, 6-bromoquinolin-4-one
PubChem CID 12403680
IUPAC Name 6-bromo-1H-quinolin-4-one
SMILES C1=CC2=C(C=C1Br)C(=O)C=CN2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B48991G
View Documents
TCI America
B48991G
1 g
Each for $119.36
Only null left
 
Description

Description

Specifications

Chemical Identifiers

145369-94-4
224.057
XKLBNOHKHRAXKK-UHFFFAOYSA-N
12403680
C1=CC2=C(C=C1Br)C(=O)C=CN2
C9H6BrNO
MFCD00272435
6-bromoquinolin-4-ol, 6-bromo-4-hydroxyquinoline, 6-bromoquinolin-4 1h-one, 6-bromo-quinolin-4-ol, 4-quinolinol, 6-bromo, 6-bromo-1,4-dihydroquinolin-4-one, 6-bromo-4-quinolinol, 4-hydroxy-6-bromoquinoline, 6-bromo-4-hyrdroxyquinoline, 6-bromoquinolin-4-one
6-bromo-1H-quinolin-4-one

Specifications

145369-94-4
C9H6BrNO
1 g
XKLBNOHKHRAXKK-UHFFFAOYSA-N
6-bromo-1H-quinolin-4-one
12403680
≥98.0% (GC,T)
6-Bromo-4-hydroxyquinoline
283°C
MFCD00272435
6-bromoquinolin-4-ol, 6-bromo-4-hydroxyquinoline, 6-bromoquinolin-4 1h-one, 6-bromo-quinolin-4-ol, 4-quinolinol, 6-bromo, 6-bromo-1,4-dihydroquinolin-4-one, 6-bromo-4-quinolinol, 4-hydroxy-6-bromoquinoline, 6-bromo-4-hyrdroxyquinoline, 6-bromoquinolin-4-one
C1=CC2=C(C=C1Br)C(=O)C=CN2
224.057
224.06
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents