missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-4-hydroxyquinoline 98.0+%, TCI America™

Catalog No. B48991G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B48991G Supplier TCI America Supplier No. B48991G
Only null left

Chemical Identifiers

CAS 145369-94-4
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD00272435
InChI Key XKLBNOHKHRAXKK-UHFFFAOYSA-N
Synonym 6-bromoquinolin-4-ol, 6-bromo-4-hydroxyquinoline, 6-bromoquinolin-4 1h-one, 6-bromo-quinolin-4-ol, 4-quinolinol, 6-bromo, 6-bromo-1,4-dihydroquinolin-4-one, 6-bromo-4-quinolinol, 4-hydroxy-6-bromoquinoline, 6-bromo-4-hyrdroxyquinoline, 6-bromoquinolin-4-one
PubChem CID 12403680
IUPAC Name 6-bromo-1H-quinolin-4-one
SMILES C1=CC2=C(C=C1Br)C(=O)C=CN2

Specifications

Melting Point 283°C
Quantity 1 g
Formula Weight 224.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-4-hydroxyquinoline