missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-pyridinemethanol 98.0+%, TCI America™

Catalog No. B34941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34941G 1 g
B34945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34941G Supplier TCI America Supplier No. B34941G

Chemical Identifiers

CAS 122306-01-8
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD04974508
InChI Key QPPDKOIDAYZUHN-UHFFFAOYSA-N
Synonym 6-bromopyridin-3-yl methanol, 2-bromo-5-hydroxymethyl pyridine, 6-bromo-pyridin-3-yl-methanol, 6-bromo-3-pyridinemethanol, 6-bromo-pyridin-3-yl methanol, 2-bromo-pyridin-5-yl-methanol, 6-bromo-3-pyridyl methanol, 2-bromo-5-hydroxymethylpyridine, 6-bromo-3-pyridinyl methanol, 6-bromopyridine-3-methanol
PubChem CID 6421249
IUPAC Name (6-bromopyridin-3-yl)methanol
SMILES C1=CC(=NC=C1CO)Br

Specifications

Melting Point 50°C
Quantity 1 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-3-pyridinemethanol