missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. B34911G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34911G 1 g
B34915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34911G Supplier TCI America Supplier No. B34911G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 149806-06-4
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.01
MDL Number MFCD04115419
InChI Key PVUKGNBRJFTFNJ-UHFFFAOYSA-N
Synonym 6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
PubChem CID 11769234
IUPAC Name 6-bromopyridine-3-carbaldehyde
SMILES BrC1=CC=C(C=O)C=N1

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 1 g
Formula Weight 186.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-3-pyridinecarboxaldehyde