missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-pyridinecarboxaldehyde, 95%, Thermo Scientific™

Chemical Identifiers

CAS 149806-06-4
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.01
MDL Number MFCD04115419
InChI Key PVUKGNBRJFTFNJ-UHFFFAOYSA-N
Synonym 6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
PubChem CID 11769234
IUPAC Name 6-bromopyridine-3-carbaldehyde
SMILES BrC1=CC=C(C=O)C=N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC399850010
SDS View Documents
Thermo Scientific Chemicals
399850010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399850050
SDS View Documents
Thermo Scientific Chemicals
399850050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399850250
SDS View Documents
Thermo Scientific Chemicals
399850250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

149806-06-4
186.01
PVUKGNBRJFTFNJ-UHFFFAOYSA-N
11769234
BrC1=CC=C(C=O)C=N1
C6H4BrNO
MFCD04115419
6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
6-bromopyridine-3-carbaldehyde

Specifications

149806-06-4
Glass bottle
MFCD04115419
6-bromonicotinaldehyde, 2-bromo-5-formylpyridine, 2-bromopyridine-5-carbaldehyde, 6-bromo-3-pyridinecarboxaldehyde, 2-bromopyridine-5-carboxaldehyde, 3-pyridinecarboxaldehyde, 6-bromo, 6-bromo-pyridine-3-carbaldehyde, 6-bromopyridine-3-carboxaldehyde, 2-bromo-5-pyridinecarbaldehyde, 6-bromo-3-formylpyridine
BrC1=CC=C(C=O)C=N1
186.01
186
6-Bromo-3-pyridinecarboxaldehyde
94% min. (GC)
C6H4BrNO
1g
PVUKGNBRJFTFNJ-UHFFFAOYSA-N
6-bromopyridine-3-carbaldehyde
11769234
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only