missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Bromo-3-formylchromone 97.0+%, TCI America™
$78.29 - $275.87
Chemical Identifiers
| CAS | 52817-12-6 |
|---|---|
| Molecular Formula | C10H5BrO3 |
| Molecular Weight (g/mol) | 253.05 |
| MDL Number | MFCD00191849 |
| InChI Key | PCEZXSJBHMOQFT-UHFFFAOYSA-N |
| PubChem CID | 614309 |
| IUPAC Name | 6-bromo-4-oxo-4H-chromene-3-carbaldehyde |
| SMILES | BrC1=CC=C2OC=C(C=O)C(=O)C2=C1 |
Chemical Identifiers
| 52817-12-6 | |
| 253.05 | |
| PCEZXSJBHMOQFT-UHFFFAOYSA-N | |
| 6-bromo-4-oxo-4H-chromene-3-carbaldehyde |
| C10H5BrO3 | |
| MFCD00191849 | |
| 614309 | |
| BrC1=CC=C2OC=C(C=O)C(=O)C2=C1 |
Specifications
| 52817-12-6 | |
| C10H5BrO3 | |
| 5 g | |
| PCEZXSJBHMOQFT-UHFFFAOYSA-N | |
| 6-bromo-4-oxo-4H-chromene-3-carbaldehyde | |
| 614309 | |
| ≥97.0% (GC) | |
| 6-Bromo-3-formylchromone |
| 191°C | |
| MFCD00191849 | |
| 6-bromo-3-formylchromone, 6-bromo-4-oxo-4h-chromene-3-carbaldehyde, chembl85756, 6-bromochromone-3-carboxaldehyde, 6-bromo-4-oxo-4h-1-benzopyran-3-carboxaldehyde, 4h-1-benzopyran-3-carboxaldehyde, 6-bromo-4-oxo, 6-bromo-4-oxo-chromene-3-carbaldehyde, 4h-1-benzopyran-3-carboxaldehyde,6-bromo-4-oxo, acmc-209l1y, 6-bromochromone-3-carbaldehyde | |
| BrC1=CC=C2OC=C(C=O)C(=O)C2=C1 | |
| 253.05 | |
| 253.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : No