missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-3-formylchromone, 97%

Catalog No. AAH3205303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3205303 1 g
AAH3205306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3205303 Supplier Thermo Scientific Chemicals Supplier No. H3205303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52817-12-6 | C10H5BrO3 | 253.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52817-12-6
Molecular Formula C10H5BrO3
Molecular Weight (g/mol) 253.05
MDL Number MFCD00191849
InChI Key PCEZXSJBHMOQFT-UHFFFAOYSA-N
PubChem CID 614309
IUPAC Name 6-bromo-4-oxochromene-3-carbaldehyde
SMILES BrC1=CC=C2OC=C(C=O)C(=O)C2=C1

Specifications

Melting Point 190°C to 193°C
Quantity 1 g
Synonym 6-bromo-3-formylchromone, 6-bromo-4-oxo-4h-chromene-3-carbaldehyde, chembl85756, 6-bromochromone-3-carboxaldehyde, 6-bromo-4-oxo-4h-1-benzopyran-3-carboxaldehyde, 4h-1-benzopyran-3-carboxaldehyde, 6-bromo-4-oxo, 6-bromo-4-oxo-chromene-3-carbaldehyde, 4h-1-benzopyran-3-carboxaldehyde,6-bromo-4-oxo, acmc-209l1y, 6-bromochromone-3-carbaldehyde
Formula Weight 253.05
Percent Purity 97%
Chemical Name or Material 6-Bromo-3-formylchromone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only